Learn More
Simport™ Scientific Swingsette™ Biopsy Cassettes In Quickload™ Sleeves
Shop All Simport Scientific ProductsDescription
- Made of acetal
- 1 mm square openings to maximize fluid exchange
- Sleeved cassettes for use with ThermoFisher printers
- Anterior writing area at a 45° angle
- Large tab for convenient and easy opening
- Covers packaged separately
- Easy-to-assemble disposable cover with special hinge
Specifications
Specifications
| Color | Tan |
| Product Type | Biopsy Cassette |