Learn More
Simport™ Scientific Swingsette™ Biopsy Cassettes In Quickload™ Sleeves
Catalog No. p-4758085
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Suitable for hoppers accepting plastic sleeves, these Swingsette™ Biopsy Cassettes In Quickload™ Sleeves will load in cassette labeling instruments in one simple operation. Specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets.
  • Made of acetal
  • 1 mm square openings to maximize fluid exchange
  • Sleeved cassettes for use with ThermoFisher printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Specifications
Color White
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.