Learn More
Simport™ Scientific Swingsette™ Biopsy Processing/Embedding Cassettes
Catalog No. 22038703 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
White
Yellow
Closure Type:
Covers Packaged Separately
Removable Cover
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Closure Type
22-038-703 Orange Covers Packaged Separately
22-038-670 Pink Removable Cover
22-038-672 Yellow Removable Cover
22-038-673 Blue Removable Cover
22-038-674 Peach Removable Cover
22-038-680 Aqua Removable Cover
22-038-678 Lilac Removable Cover
22-038-692 White Covers Packaged Separately
22-038-694 Green Covers Packaged Separately
22-038-693 Pink Covers Packaged Separately
22-038-695 Yellow Covers Packaged Separately
22-038-697 Peach Covers Packaged Separately
22-038-696 Blue Covers Packaged Separately
22-038-699 Gray Covers Packaged Separately
22-038-702 Lilac Covers Packaged Separately
22-038-704 Aqua Covers Packaged Separately
22-038-671 Green Removable Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Catalog No. 22-038-703 Supplier Simport™ Scientific Supplier No. M51811
Add to Cart
Add to Cart
This item is not returnable. View return policy
These Swingsette™ Biopsy Cassettes are specially designed to hold biopsy specimens during the processing/embedding process as well as in storage cabinets. Molded from acetal, they keep specimens safely submerged in liquid and are resistant to the chemical action of most histological solvents.
  • Made of acetal
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Easy-to-remove pre-attached cover with special hinge
  • Can be opened and closed as often as necessary
  • 1 mm square openings to maximize fluid exchange
Specifications
Packaging Type Covers Packaged Separately
Closure Type Covers Packaged Separately
Color Orange
Material High Density Polymer
Product Type Biopsy Cassette