Learn More
Simport™ Scientific Swingsette™ Tissue Cassettes In Quickload™ Sleeves
Shop All Simport Scientific ProductsDescription
- Made of acetal
- Sleeved cassettes for use with Thermo Fisher printers
- Anterior writing area at a 45° angle
- Large tab for convenient and easy opening
- Covers packaged separately
- Easy-to-assemble disposable cover with special hinge
Specifications
Specifications
| Closure Included | Yes |
| Color | Yellow |
| Material | Acetal Polymer |
| Product Type | Tissue Cassette |