Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes
Shop All Simport Scientific ProductsDescription
- Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
- Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
- Cover can be removed and re-inserted easily without danger of specimen loss
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Covers packaged separately
Specifications
Specifications
| Closure Included | No |
| Color | Gray |
| Material | High Density Polymer |
| Product Type | Tissue Cassette |