Learn More
Simport™ Scientific Swingsette™ Tissue Processing/Embedding Cassettes
Catalog No. 22038685 Shop All Simport Scientific Products
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-038-685 Aqua
22-038-681 White
22-038-682 Gray
22-038-683 Lilac
22-038-684 Orange
22-038-688 Yellow
22-038-687 Green
22-038-686 Pink
22-038-689 Blue
22-038-691 Tan
22-038-690 Peach
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 22-038-685 Supplier Simport™ Scientific Supplier No. M51712
Add to Cart
Add to Cart
Disposable plastic Swingsette™ Tissue Processing/Embedding Cassettes hold tissue specimens during processing, embedding and storage.
  • Molded from a special high-density polymer, the cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents
  • Special hinge holds base and cover together, allowing cassettes to be opened and closed repeatedly
  • Cover can be removed and re-inserted easily without danger of specimen loss
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • Covers packaged separately
Specifications
Closure Included No
Color Aqua
Material High Density Polymer
Product Type Tissue Cassette