Learn More
Simport™ Scientific T100 BIOTUBE™ Tube Racks
Click to view available options
Color:
Blue
Green
Red
Yellow
Sterility:
Non-sterile
Sterile
Includes:
12 Strips of 8 Tubes
96 Plain Individual Tubes
Description
- Stands on four legs and can be removed from the base of the box and placed on a lab counter as a self-standing support
- Can be placed in a refrigerator or freezer shelf for improved air circulation around tubes, or in a water bath to allow controlled warming of the tubes and their contents
- Standard 96-well on center spacing of tubes
- Color coding system using colored interchangeable plastic grids
- Easy to read numbers and letters used on the box cover for sample identification
- Also ideal for storing, freezing and transporting reagents and specimens
- Availaible in red, blue, green and yellow
Specifications
Specifications
Includes | 96 Plain Individual Tubes |
Product Type | T100 BIOTUBE Tube Rack |
Color | Yellow |
Holds | Microtubes (1.2 mL) |
Material | Polypropylene |
Positions | 96 |
Sterility | Sterile |
Quantity | 10/Cs. |