Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific T100 BIOTUBE™ Caps

Catalog No. 0550023 Shop All Simport Scientific Products
Change view
Click to view available options
Sterility:
Sterile
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
05-500-23 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 05-500-23 Supplier Simport™ Scientific Supplier No. T10035
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

T100 BIOTUBE™ Caps are made of polyethylene and are not autoclavable. Tubes with a cap in place will hold 1.1 mL. They are available individually and also in strips of 8 or 12.

  • Polypropylene construction
  • Caps sold separately
  • Gross volume: 1.2 mL, with cap : 1.1 mL
  • Used for procedures requiring low surface tension such as protein, nucleic acid work
  • Designed to avoid potentially harmful lubricants while minimizing liquid retention
  • Applications: Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA

Specifications

Autoclavable Not Autoclavable
Capacity (Metric) 1.2 mL
Color Clear
Material Polyethylene
For Use With (Application) Ideal for HTLV-III testing, bacterial and hybridoma cell uptake studies, cell harvesting, pharmaceutical quality control, receptor binding assays, RIA and EIA
For Use With (Equipment) T100 Biotube
Includes Strips of 8 Plug Caps
No. per Pack 600/Cs.
Sterility Sterile
Closure Type Plug
Max. RCF 2000 x g
Product Type Plug Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.