Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simport™ Scientific T336 Micrewtube Microcentrifuge Tubes

Catalog No. 05500103 Shop All Simport Scientific Products
Change view
Click to view available options
Capacity (Metric):
1.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
05-500-103 1.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-500-103 Supplier Simport™ Scientific Supplier No. T3365
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Cap Style Pre-attached Loop
Certifications/Compliance RNase, DNase, DNA, pyrogen-free
Autoclavable Not Autoclavable
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
For Use With (Application) Ideal for freezer storage, boiling application, centrifugation etc. and will fit most standard microcentrifuge rotors
Sterility Non-sterile
Bottom Shape Conical
Max. RCF 20,000 x g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.