Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Simulaids™ Rescue Randy Manikin



Click to view available options
:
Rescue Randy Adult Manikin
Description
- Can be safely used in situations too hazardous or uncomfortable for human volunteers
- Full-bodied manikin made of durable vinyl with 4100 lb. test plastic-coated cables
- Features include articulated joints, weight distribution according to human weight distribution chart
- Size: 50 x 21 x 12 in.
- Weight: 125 lb.
Order Info
Carry/storage bag available separately.
Specifications
Specifications
Product Type | Adult Rescue Randy Manikin |
For Use With (Application) | Lifelike Adult or Juvenile Victim Handling, Transportation and Extrication Training |
Includes | Articulated joints and weight distribution according to human weight distribution chart |
Material | Vinyl, Steel |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction