Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Simvastatin
GSA_VAGREENER_CHOICESDP

Catalog No. 196550 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-655-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-655-0 Supplier R&D Systems™ Supplier No. 1965/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. HMG-CoA reductase inhibitor

HMG-CoA reductase inhibitor; decreases levels of low density lipoprotein. Has multiple biological effects including bone formation stimulation, inhibition of smooth muscle cell proliferation and migration, induction of ferroptosis, and anticancer and anti-inflammatory activity. Inactive lactone prodrug of simvastatin hydroxy acid, naturally bioactivated in vivo following oral administration.

Deuterated analog also available.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 79902-63-9
Molecular Formula C25H38O5

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 418.57
Purity >98%
Molecular Weight (g/mol) 418.574
Chemical Name or Material Simvastatin
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.