Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simvastatin, 98%

Catalog No. p-7077758
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 79902-63-9 | C25H38O5 | 418.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79902-63-9
Molecular Formula C25H38O5
Molecular Weight (g/mol) 418.57
InChI Key RYMZZMVNJRMUDD-HGQWONQESA-N
Synonym simvastatin, zocor, synvinolin, sinvacor, denan, lipex, simvastatine, sivastin, lodales, simvastatina
PubChem CID 54454
ChEBI CHEBI:9150
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
SMILES CCC(C)(C)C(=O)OC1CC(C=C2C1C(C(C=C2)C)CCC3CC(CC(=O)O3)O)C

Specifications

Melting Point 127.0°C to 132.0°C
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 418.57
Percent Purity 98%
Chemical Name or Material Simvastatin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
Obtain special instructions before use.
IF exposed or concerned:

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.