Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SIN-1 chloride
GREENER_CHOICESDP

Catalog No. 075650 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-565-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-565-0 Supplier R&D Systems™ Supplier No. 0756/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Water-soluble NO donor

This compound (the active product of the prodrug SIN-10, molsidomine) acts as a vasodilator and inhibitor of platelet aggregation; longer acting than nitroprusside or nitroglycerin. Decreases myocardial necrosis and reperfusion-induced endothelial dysfunction in models of myocardial ischemia-reperfusion.

Chemical Identifiers

CAS 16142-27-1
Molecular Formula C6H11ClN4O2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 206.63
Purity >98%
Chemical Name or Material SIN-1 chloride
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.