Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIN-1 hydrochloride, 97%, Thermo Scientific™

Catalog No. AC328850100
Click to view available options
Quantity:
10 mg
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16142-27-1
Molecular Formula C6H10N4O2·HCl
MDL Number MFCD00221615
Synonym 3-Morpholinosydnonimine hydrochloride

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 mg
Formula Weight 206.66
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material SIN-1 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.