Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody
Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody
Supplier: Thermo Scientific PA572794
Description
Sin Nombre Virus Glycoprotein 1 Polyclonal Antibody for ELISA

Specifications
Sin Nombre Virus Glycoprotein 1 | |
Polyclonal | |
Unconjugated | |
SNVsMgp1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2654026 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
SNVsMgp1 | |
Sin Nombre virus glycoprotein 1 antibody for a 19 amino acid peptide near the amino terminus of the Sin Nombre virus glycoprotein. The immunogen is within amino acids 60 - 110 of Sin Nombre Virus Glycoprotein 1. | |
100 μg | |
Primary | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction