Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Sin Nombre Virus Nucleocapsid Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572796
Description
Sin Nombre Virus Nucleocapsid Polyclonal Antibody for ELISA
Sin Nombre Virus Nucleocapsid is part of the internal capsid protein structure of the Sin Nombre virus (SNV) which is a rodent-borne hantavirus of the family Bunyaviridae, an enveloped, negative-sense RNA viruses with a tripartite genome that can cause hantavirus pulmonary syndrome (HPS). Hantavirus glycoprotein precursor (GPC) is posttranslationally cleaved into two glycoproteins G1 (Gn) and G2 (Gc). The G1 glycoprotein is is thought to be degraded by the host autophagy machinery, and this autophagic clearance is required for efficient virus repliaction.
Specifications
Sin Nombre Virus Nucleocapsid | |
Polyclonal | |
Unconjugated | |
SNVsSgp1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2943142 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
SNVsSgp1 | |
Sin Nombre virus nucleocapsid antibody for a 19 amino acid peptide near the center of the Sin Nombre virus nucleocapsid. The immunogen is within amino acids 170 - 220 of Sin Nombre Virus Nucleocapsid. | |
100 μg | |
Primary | |
Virus | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction