Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sin1/MAPKAP1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11040424AF700
Description
Sin1/MAPKAP1 Polyclonal antibody specifically detects Sin1/MAPKAP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Sin1/MAPKAP1 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
JC310, MEKK2-interacting protein 1, MGC2745, MIP1SIN1SIN1b, Mitogen-activated protein kinase 2-associated protein 1, mitogen-activated protein kinase associated protein 1, mSIN1, ras inhibitor MGC2745, SAPK-interacting protein 1, SIN1g, stress-activated map kinase interacting protein 1, Stress-activated map kinase-interacting protein 1, stress-activated protein kinase-interacting 1, target of rapamycin complex 2 subunit MAPKAP1, TORC2 subunit MAPKAP1 | |
The immunogen recognized by this antibody maps to a region between residue 470 and the C-terminus (residue 522) of human stress-activated map kinase interacting protein 1 using the numbering given in entry NP_001006618.1 (GeneID 79109). | |
0.1 mL | |
Autophagy, Cancer, mTOR Pathway, Protein Kinase | |
79109 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction