Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIN3A Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298901R
Description
SIN3A Polyclonal specifically detects SIN3A in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SIN3A | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of mouse SIN3A. | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
25942 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction