Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIN3A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP238564
Description
SIN3A Polyclonal specifically detects SIN3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
SIN3A | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Q96ST3 | |
SIN3A | |
This antibody was developed against a recombinant protein corresponding to amino acids: SDDPVEAERWSDYVERYMNSDTTSPELREHLAQKPVFLPRNLRRIRKCQRGREQQEKEGKEGNSKKTMENVDSLDKLECRFKLNSYKMVYVIKSEDYMYRRTALLRAHQSHERVSK | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
DKFZp434K2235, FLJ90319, Histone deacetylase complex subunit Sin3a, KIAA0700, paired amphipathic helix protein Sin3a, SIN3 homolog A, transcription regulator (yeast), SIN3 homolog A, transcriptional regulator (yeast), transcriptional co-repressor Sin3A, Transcriptional corepressor Sin3a, transcriptional regulator, SIN3A | |
Rabbit | |
Affinity Purified | |
RUO | |
25942 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction