Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIN3B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP220368
Description
SIN3B Polyclonal specifically detects SIN3B in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
SIN3B | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
Histone deacetylase complex subunit Sin3b, KIAA0700paired amphipathic helix protein Sin3b, SIN3 homolog B, transcription regulator (yeast), SIN3 homolog B, transcriptional regulator, SIN3 homolog B, transcriptional regulator (yeast), Transcriptional corepressor Sin3b | |
Rabbit | |
40 kDa | |
0.1 mL | |
Primary | |
Human, Mouse. Species reactivity based on sequence identity: Rat (91%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
0.57mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500 | |
O75182 | |
SIN3B | |
Recombinant protein encompassing a sequence within the center region of human SIN3B. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
23309 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction