Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIN3B Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA584001
Description
Immunogen sequence: LISYYVKRQP AIQKEDQGTI HQLLHQFVPS LFFSQQLDLG ASEESADEDR DSPQGQTTDP SERKKPAPGP HSSPPEEKGA FGDAPATEQP P.
Acts as a transcriptional repressor. Interacts with MXI1 to repress MYC responsive genes and antagonize MYC oncogenic activities. Interacts with MAD-MAX heterodimers by binding to MAD. The heterodimer then represses transcription by tethering SIN3B to DNA. Also forms a complex with FOXK1 which represses transcription.
Specifications
SIN3B | |
Polyclonal | |
Unconjugated | |
SIN3B | |
2810430C10Rik; Histone deacetylase complex subunit Sin3b; KIAA0700; Paired amphipathic helix protein Sin3b; SIN3 homolog B, transcription regulator; SIN3 homolog B, transcriptional regulator; SIN3 transcription regulator family member B; SIN3 transcription regulator homolog B; SIN3B; Transcriptional corepressor Sin3b; transcriptional regulator, SIN3 yeast homolog B; transcriptional regulator, SIN3B; transcriptional regulator, SIN3B (yeast) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75182 | |
SIN3B | |
Recombinant protein corresponding to Human SIN3B. Recombinant protein control fragment (Product #RP-98415). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction