Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Single Battery Holder
GREENER_CHOICESCI_ED

Catalog No. S01279 Shop All Supertek Scientific Products
Click to view available options
For Use With (Equipment):
C-cell Battery
D-cell Battery

Catalog No. S01279

Supplier: Supertek™ PH60112AS3

Only null left
Add to Cart
Add to Cart

Single Battery Holders can be snapped together in parallel or series to extend capacity or voltage.

  • Features clips for easy connection.
  • Snap battery holders together in parallel or series to extend capacity or voltage.
  • Made of unbreakable plastic.
  • Curved sides help prevent the battery from falling out when turned upside down.
  • Ages 7 plus

Specifications

Product Type Battery Holder
Count (Outlets) Single
For Use With (Equipment) D-cell Battery
Length (English) 3.25 in.
Length (Metric) 8.3 cm
Material Plastic
Voltage 0/1 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.