Learn More
AccuTec Blades Single Edge Long Blades
AccuTec Blades Single Edge Long Blades are excellent for shaping and shaving material in a variety of industries
$133.50 - $236.50
Specifications
For Use With (Application) Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Sterility Non-sterile
View More Specs
Products 3
Catalog Number Mfr. No. Back Style Price Quantity & Availability  
Catalog Number Mfr. No. Back Style Price Quantity & Availability  
17-989-151
View Documents
AccuTec Blades
940367
Aluminum
Each for $133.50
Case of 20 EA for $2,150.00
Add to cart
17-989-149
View Documents
AccuTec Blades
940213
Stainless Steel
Each for $236.50
Case of 20 EA for $3,790.00
Add to cart
17-989-150
View Documents
AccuTec Blades
940314
Unbacked
Each for $166.50
Add to cart
Description
Description
  • 3 Facet Blades
  • .009 in. Stainless Steel
  • Coated, or uncoated options
  • Offered unwrapped, or individually wrapped
  • Fits standard prep razor handle
Specifications
Specifications
Cutting Catheter Tubing, Scraping, Manufacturing, Industrial and Medical Applications
Non-sterile