Learn More
Eisco™ Single Sockets
SCI_ED
Catalog No. S89356 Shop All Eisco Products
Change view
Click to view available options
Standard Taper Joint No.:
10/19
12/21
14/23
34/35
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint No.
S89356 10/19
S89357 12/21
S89358 14/23
S89362 34/35
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S89356 Supplier Eisco™ Supplier No. CH0823A
Only null left
Add to Cart
Add to Cart
A variety of standard socket sizes are available, each with a minimum shank length of 100 mm
  • Single ground glass sockets for connecting glassware
  • Made from high quality borosilicate 3.3 glass
  • Autoclavable
  • Warranty and Services
    1 year limited manufacturer warranty
    Specifications
    For Use With (Application) Connect glassware
    Height (Metric) 12.7 cm
    Length (Metric) 3 cm
    Material Borosilicate Glass (3.3)
    Product Type Single Socket
    Quantity 1
    Standard Taper Joint No. 10/19