Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIPA1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP118917AF532
Description
SIPA1 Polyclonal antibody specifically detects SIPA1 in Human, Mouse samples. It is validated for Western BlotSpecifications
SIPA1 | |
Polyclonal | |
Western Blot | |
GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1 | |
The immunogen recognized by this antibody maps to a region between residue 992 to 1042 of human signal-induced proliferation-associated protein 1 using the numbering given in entry NP_006738.3 (GeneID 6494). | |
0.1 mL | |
Apoptosis, Breast Cancer | |
6494 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction