Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIPA1 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298740PCP
Description
SIPA1 Polyclonal antibody specifically detects SIPA1 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SIPA1 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
GTPase-activating protein Spa-1, MGC102688, MGC17037, p130 SPA-1, signal-induced proliferation-associated 1, signal-induced proliferation-associated protein 1, sipa-1, SPA1signal-induced proliferation-associated gene 1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SIPA1. | |
0.1 mL | |
Apoptosis, Breast Cancer | |
6494 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction