Learn More
Invitrogen™ SIPA1 Polyclonal Antibody

Description
Immunogen sequence: ESGRPRRSFS ELYTLSLQEP SRRGAPDPVQ DEVQGVTLLP TTKQLLHLCL QDGGSPPGPG DLAEERTEFL HSQNSLSPRS SLSDEAPVLP NTTPDLLLAT TAKPSVPSAD SETPLTQDRP GSPSGSEDKG NPAPELRASF LPRTLSLRNS ISRIMSEAGS GTLEDEWQAI SEIASTCNTI LESLSREGQP IPESGDPKGT PKSDAEPEPG NLSEKVSHLE SMLRKLQEDL QKEKADRAAL EEEVRSLRHN NRRLQAESES AATRLLLASK QLGSPTADLA; Positive Samples: Raji, K-562, HeLa, LO2, Mouse spleen; Cellular Location: Cytoplasm, Endomembrane system, Nucleus, Peripheral membrane protein, perinuclear region.
Specifications
Specifications
| Antigen | SIPA1 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.53 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | SIPA1 |
| Gene Accession No. | P46062, Q96FS4 |
| Gene Alias | GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1 |
| Gene Symbols | SIPA1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.