Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558957 Supplier Invitrogen™ Supplier No. PA558957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPSGSEDKGN PAPELRASFL PRTLSLRNSI SRIMSEAGSG TLEDEWQAIS EIASTCNTIL ESLSREGQPI PESGDPKGTP KSDAEPEPGN LSEKVSHLES M Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIPA1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SIPA1
Gene Accession No. Q96FS4
Gene Alias GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1
Gene Symbols SIPA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SIPA1. Recombinant protein control fragment (Product #RP-97563).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6494
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.