Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987241
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7241 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7241 Supplier Bioss Supplier No. BS6399R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The product of this gene is a mitogen induced GTPase activating protein. It exhibits a specific GAP activity for Ras-related regulatory proteins Rap1 and Rap2, but not for Ran or other small GTPases. This protein may also hamper mitogen-induced cell cycle progression when abnormally or prematurely expressed. It is localized to the perinuclear region. Two alternatively spliced variants encoding the same isoform have been characterized to date.

Specifications

Antigen SIPA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIPA1
Gene Accession No. Q96FS4
Gene Alias GTPase-activating protein Spa-1; MGC102688; MGC17037; p130 SPA-1; signal-induced proliferation associated gene 1; signal-induced proliferation-associated 1; signal-induced proliferation-associated gene 1; Signal-induced proliferation-associated protein 1; SIPA1; Sipa-1; SPA1
Gene Symbols SIPA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SIPA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6494
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.