Learn More
Invitrogen™ SIPA1L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551761
Description
Immunogen sequence: IRQRSNSDIT ISELDVDSFD ECISPTYKTG PSLHREYGST SSIDKQGTSG ESFFDLLKGY KDDKSDRGPT PTKLSDFLIT GGGKGSGFSL DVIDGPISQR ENLRLFKERE KPLKRRSKSE TGDSSIFRKL RNAKGEELGK SSDLEDNRSE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
Signal-induced proliferation associated-like protein 1 (SIPA1L1) is a member of the SIPA1 family of RapGAPs. SIPA1L1 was initially identified as a binding partner and degradation target of the E6 oncoprotein of high-risk papillomaviruses. Recently, it was discovered that Casein kinase I epsilon (CKIe), a Wnt-regulated kinase that regulates Wnt/b-catenin signaling, also can bind to the carboxy-terminus of SIPA1L1. CKIe phosphorylates SIPA1L1, thereby reducing its stability and alleviating its inhibition of Rap1, a protein required for Wnt8/CKIe-mediated gastrulation during embryogenesis, suggesting SIPA1L1 plays important roles in embryo development as well as control of cell proliferation.
Specifications
SIPA1L1 | |
Polyclonal | |
Unconjugated | |
SIPA1L1 | |
4931426N11Rik; AW213287; E6-targeted protein 1; E6TP1; H_DJ1140G11.1; High-risk human papilloma viruses E6 oncoproteins targeted protein 1; Kiaa0440; mKIAA0440; signal induced proliferation associated 1 like 1; signal-induced proliferation-associated 1 like 1; signal-induced proliferation-associated 1-like protein 1; SIPA1L1; SIPA1-like protein 1; Spa1; Spa-1; SPA-1 like protein p1294; SPA-1-like protein p1294; SPAL; Spar; Spine-associated Rap GTPase-activating protein; spine-associated Rap guanosine triphosphatase (GTPase) activating protein (GAP); WUGSC:H_DJ1140G11.1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O43166 | |
SIPA1L1 | |
Recombinant protein corresponding to Human SIPA1L1. Recombinant protein control fragment (Product #RP-89050). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.