Learn More
SIPA1L2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304677A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.Specifications
SIPA1L2 | |
Polyclonal | |
Unconjugated | |
SIPA1L2 | |
SPAL2; SIPA1-like protein 2; SPA-1-like 2; signal-induced proliferation-associated 1-like protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57568 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9P2F8 | |
SIPA1L2 | |
Between 1250 and 1300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.