Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIPA1L2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988132
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8132 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8132 Supplier Bioss Supplier No. BS7927R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Signal-induced proliferation associated-like protein 2 (SIPA1L2) is a member of the SIPA1 family of RapGAPs. Little is known of the role of the SIPA1L2 protein, but recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is likely that SIPA1L2 plays a role in embryo development and cell proliferation, possibly including oncogenesis.

Specifications

Antigen SIPA1L2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SIPA1L2
Gene Accession No. Q9P2F8
Gene Alias BC058408; Kiaa0545; KIAA1389; mKIAA1389; serine-rich synapse associated protein 2; Serine-rich synapse-associated protein; Sersap2; signal induced proliferation associated 1 like 2; signal-induced proliferation-associated 1 like 2; Signal-induced proliferation-associated 1-like protein 2; SIPA1L2; SIPA1-like protein 2; SPA-1-like 2; SPAL2; Spar2
Gene Symbols SIPA1L2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SIPA1L2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57568
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.