Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SIPA1L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520849
Description
A suggested positive control is SK-N-SH cell lysate. PA5-20849 can be used with blocking peptide PEP-0963.
Signal-induced proliferation associated-like protein 2 (SIPA1L3) is a member of the SIPA1 family of RapGAPs. SIPA1L3 was cloned as a novel molecular component of tight junctions and adherens junctions, suggesting that may function to balance Rap1 by counteracting RapGEFs. Recent studies of SIPA indicate that its deregulation can cause myeloproliferative stem cell disorders in mice and increased metastases in human cancers. Other studies suggest SIPA1L1 may play important roles in embryo development and control of cell proliferation. Based on the amount of homology between SIPA family members, it is possible that SIPA1L3 plays other roles development and cell proliferation.
Specifications
SIPA1L3 | |
Polyclonal | |
Unconjugated | |
SIPA1L3 | |
2610511M17Rik; CTRCT45; KIAA0545; signal induced proliferation associated 1 like 3; signal-induced proliferation-associated 1 like 3; signal-induced proliferation-associated 1-like protein 3; Sipa1l3; SIPA1-like protein 3; SPA-1-like 3; SPA-1-like protein 3; Spal3; SPAR3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23094, 292771, 74206 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
G3X9J0, O60292 | |
SIPA1L3 | |
A 15 amino acid peptide from near the amino terminus of human SIPA1L3. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction