Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIPA1L3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP317768100UL
This item is not returnable.
View return policy
Description
SIPA1L3 Polyclonal antibody specifically detects SIPA1L3 in Human samples. It is validated for ImmunofluorescenceSpecifications
SIPA1L3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
KIAA0545, Signal-Induced Proliferation-Associated 1 Like 3, Signal-Induced Proliferation-Associated 1-Like Protein 3, SIPA1-Like Protein 3, SPA-1-Like 3, SPA-1-Like Protein 3, SPAL3 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: QPSGSFSTPGSATYVRYKPSPERYTAAPHPLLSLDPHFSHDGTSSGDSSSGGLTSQESTMERQKPEPLWHVPAQARLSAIAGSSGNKHPSR | |
100 μg | |
Cancer | |
23094 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunofluorescence | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction