Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP alpha/CD172a Antibody (602411), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4546L
Description
SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
| SIRP alpha/CD172a | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 602411 | |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
| Chinese hamster ovary cell line CHO-derived recombinant human SIRP alpha/CD172a, Gly27-Asn370 (predicted), Accession # P78324 | |
| 0.1 mL | |
| GPCR, Protein Phosphatase, Signal Transduction | |
| 140885 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction