Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP alpha/CD172a Antibody (OX-41), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065530PE
Description
SIRP alpha/CD172a Monoclonal antibody specifically detects SIRP alpha/CD172a in Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
SIRP alpha/CD172a | |
Monoclonal | |
PE | |
Bit, BITbrain-immunoglobulin-like molecule with tyrosine-based activation motifs, Brain Ig-like molecule with tyrosine-based activation motifs, CD172 antigen-like family member A, CD172a, CD172a antigen, Inhibitory receptor SHPS-1, Macrophage fusion receptor, MFRtyrosine phosphatase SHP substrate 1, MYD1, MYD-1, MyD-1 antigen, P84, protein tyrosine phosphatase, non-receptor type substrate 1, PTPNS1, SHP substrate 1, SHPS-1, SHPS1CD172A, signal-regulatory protein alpha, Signal-regulatory protein alpha-1, Signal-regulatory protein alpha-2, Signal-regulatory protein alpha-3, SIRPalpha, SIRP-ALPHA-1, Sirp-alpha-2, SIRPalpha2, Sirp-alpha-3, SIRPtyrosine-protein phosphatase non-receptor type substrate 1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-65530 recognizes rat CD172a, also known as Signal regulatory protein (SIRP). CD172a is expressed selectively by myeloid cells and by neurons. MRC OX-41 binds to a different epitope on SIRP than ED9. It does not block the interaction of CD172a - CD47. | |
Store at 4C in the dark. | |
IgG2a |
Flow Cytometry | |
OX-41 | |
Flow Cytometry | |
SIRPA | |
Rat peritoneal macrophages | |
0.25 mL | |
GPCR, Protein Phosphatase, Signal Transduction | |
140885 | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction