Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha Mouse anti-Human, Clone: SAF4.2, MilliporeSigma™

Catalog No. MABS216325 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS216325 25 μL
MABS2163100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS216325 Supplier MilliporeSigma Supplier No. MABS216325UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Human samples. It is validated for Flow Cytometry, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP alpha
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone SAF4.2
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Human SIRP -Fc fusion protein consisting of the extracellular domain
Purification Method Protein G purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.