Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRP alpha Mouse anti-Human, Unconjugated, Clone: SAF10.1, MilliporeSigma™

Catalog No. MABS21881MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS21881MI 100 μg
MABS21882MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS21881MI Supplier MilliporeSigma Supplier No. MABS2188100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRP alpha Monoclonal antibody specifically detects SIRP alpha in Human samples. It is validated for ELISA, Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP alpha
Applications ELISA, Western Blot
Classification Monoclonal
Clone SAF10.1
Conjugate Unconjugated
Gene Accession No. NP_001035111
Gene Alias Tyrosine-protein phosphatase non-receptor type substrate 1;SHP substrate 1;SHPS-1;Brain Ig-like molecule with tyrosine-based activation motifs;Bit;CD172 antigen-like family member A;Inhibitory receptor SHPS-1;Macrophage fusion receptor;MyD-1 antigen;Signal-regulatory protein alpha-1;Sirp-alpha-1;Signal-regulatory protein alpha-2;Sirp-alpha-2;Signal-regulatory protein alpha-3;Sirp-alpha-3;p84;CD172a
Gene Symbols SIRPA, BIT, MFR, MYD1, PTPNS1, SHPS1, SIRP
Host Species Mouse
Immunogen Eukaryotically expressed SIRP-Fc fusion protein consisting of the extracellular domain.
Molecular Weight of Antigen ∼75 kDa observed; 54.97 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone SAF10.1 is a mouse monoclonal antibody that detects human Tyrosine-protein phosphatase non-receptor type substrate 1 (SIRP alpha). It targets an epitope in the membrane most distal domain (D1).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.