Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP beta 1/CD172b Antibody (010), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290729MFV450
Description
SIRP beta 1/CD172b Monoclonal antibody specifically detects SIRP beta 1/CD172b in Mouse samples. It is validated for ELISASpecifications
SIRP beta 1/CD172b | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
010 | |
ELISA | |
CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse SIRP beta 1/CD172b (Accession#: BAD26610.1; Met1-Lys363). | |
0.1 mL | |
Signal Transduction | |
10326 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction