Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP beta 1/CD172b Rabbit anti-Mouse, Clone: 10, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | SIRP beta 1/CD172b |
---|---|
Clone | 10 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
SIRP beta 1/CD172b Monoclonal specifically detects SIRP beta 1/CD172b in Mouse samples. It is validated for ELISA.Specifications
SIRP beta 1/CD172b | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD172 antigen-like family member B, CD172b, CD172b antigen, DKFZp686A05192, FLJ26614, signal-regulatory protein beta 1, signal-regulatory protein beta-1, SIRP-BETA-1, SIRP-beta-1 isoform 3 | |
Recombinant Mouse SIRP beta 1/CD172b protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
10 | |
ELISA | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
0.2 um filtered solution in PBS | |
10326 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title