Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRP beta 1/gamma Antibody (OX117), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281071DL594
Description
SIRP beta 1/gamma Monoclonal specifically detects SIRP beta 1/gamma in Human samples. It is validated for Flow Cytometry, ELISA.Specifications
SIRP beta 1/gamma | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Recombinant human SIRPgamma:CD4 fusion protein. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA | |
OX117 | |
Flow Cytometry, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
This antibody binds to human SIRP beta 1/gamma. | |
Store at 4°C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction