Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRP beta/gamma Recombinant Mouse Monoclonal Antibody (OX117)
GREENER_CHOICE

Catalog No. PIMA547959
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547959 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547959 Supplier Invitrogen™ Supplier No. MA547959
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: This antibody binds to human SIRP-beta and gamma.

SIRP beta and SIRP gamma are two members of the signal regulatory protein (SIRP) family. These proteins are transmembrane receptors that are expressed on the surface of various cells, including immune cells and neurons. The function of SIRP beta and SIRP gamma is to regulate cellular signaling pathways that are involved in immune responses, cell adhesion, and phagocytosis. Specifically, these proteins interact with the protein tyrosine phosphatase SHP-1, which is a negative regulator of immune cell activation. By binding to SHP-1, SIRP beta and SIRP gamma can inhibit immune cell activation and promote immune tolerance. In addition to their role in immune regulation, SIRP beta and SIRP gamma have also been implicated in neuronal signaling and synaptic plasticity. These proteins are thought to play a role in the formation and maintenance of synapses, as well as in the regulation of neuronal activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP beta/gamma
Applications ELISA, Flow Cytometry, Immunoprecipitation
Classification Recombinant Monoclonal
Clone OX117
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene SIRPB1
Gene Accession No. O00241, Q9P1W8
Gene Symbols SIRPB1, SIRPG
Host Species Mouse
Immunogen Recombinant human SIRPgamma:CD4 fusion protein.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10326, 55423
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.