Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRPγ (CD172g) Mouse anti-Human, PE, Clone: OX-119 (also known as OX119), BD

Catalog No. BDB566353 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566353 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566353 Supplier BD Biosciences Supplier No. 566353
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The OX-119 monoclonal antibody specifically recognizes Signal-regulatory protein gamma (SIRP-gamma or SIRP ) which is also known as CD172g or Signal-regulatory protein beta 2 (SIRP-beta-2 or SIRP 2).

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRPγ (CD172g)
Classification Monoclonal
Clone OX-119 (also known as OX119)
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias SIRPG, SIRPgamma, SIRP-gamma, SIRP beta 2, SIRP-B2, SIRPB2
Host Species Mouse
Immunogen Human SIRPgamma Recombinant Protein
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.