Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRP delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598149
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598149 Supplier Invitrogen™ Supplier No. PA598149
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SIRP delta Polyclonal Antibody for Western Blot, ELISA

SIRPD (Signal Regulatory Protein Delta) is a Protein Coding gene. An important paralog of this gene is SIRPG. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRP delta
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SIRPD
Gene Accession No. Q9H106
Gene Alias dJ576H24.4; Protein tyrosine phosphatase non-receptor type substrate 1-like 2; protein tyrosine phosphatase, non-receptor type substrate 1-like 2; PTPNS1L2; signal regulatory protein delta; signal-regulatory protein delta; SIRPD; SIRP-delta
Gene Symbols SIRPD
Host Species Rabbit
Immunogen Recombinant Human Signal-regulatory protein delta protein (30-197AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128646
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.