Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRP gamma Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521657

Catalog No. PIPA521657


Only null left
Add to Cart

Description

Description

Recommended positive controls: K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SIRP gamma (CD172g) recognizes a 55 kDa signal regulatory protein which is a member of the immunoglobulin gene superfamily. SIRP gamma is expressed on most T lymphocytes and a subset of B-cells but is absent on myeloid cells. CD172g has a truncated cytoplasmic tail, which is similar to SIRP beta, but unlike SIRP beta CD172g does not require DAP12 for expression at the cell surface. CD172g, like SIRP alpha, binds to CD47 inducing apoptosis. SIRP gamma is also involved in the negative regulation of receptor tyrosine kinase-coupled signalling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SIRP gamma
Polyclonal
Unconjugated
SIRPG
bA77C3.1; CD172 antigen-like family member B; CD172 gamma; CD172g; CD172g antigen; RP11-77C3.2; signal regulatory protein gamma; signal-regulatory protein beta 2; signal-regulatory protein beta-2; Signal-regulatory protein gamma; SIRP beta 2; SIRPB2; SIRP-B2; SIRP-beta-2; SIRPG; SIRPgamma; SIRP-gamma
Rabbit
Antigen affinity chromatography
RUO
55423
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.9 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q9P1W8
SIRPG
Recombinant protein encompassing a sequence within the Extracellular domain of human CD172 gamma. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.