Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRPB1 Monoclonal Antibody (1B1G4)
GREENER_CHOICE

Catalog No. FERMA548689
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548689 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548689 Supplier Invitrogen™ Supplier No. MA548689
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SIRPB1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow, ELISA

SIRP beta 1 is a type I transmembrane protein belonging to the SIRP family within the Ig superfamily. Members of this family are characterized by an extracellular region containing a V-set Ig domain with a J-like sequence and two C1-set Ig domains. Unlike SIRP alpha that has cytoplasmic ITIM domains, SIRP beta 1 possesses positively charged residues that allow association with ITAM motif-containing adaptor molecules. SIRP beta 1 is expressed on cells of monocyte, macrophage or dendritic lineages.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRPB1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1B1G4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SIRPB1
Gene Accession No. O00241
Gene Alias 9930027N05Rik; CD172 antigen-like family member B; CD172b; CD172b antigen; DKFZp686A05192; signal regulatory protein beta 1; signal-regulatory protein beta 1; signal-regulatory protein beta 1A; signal-regulatory protein beta-1; Signal-regulatory protein beta-1 isoform 3; SIRP BETA 1; SIRP beta 1 cell surface protein; Sirpb; SIRPB1; Sirpb1a; SIRP-beta; SIRP-beta B; SIRP-BETA-1; SIRP-beta-1 isoform 3
Gene Symbols SIRPB1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD172b (AA: extra 72-309) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10326
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.