Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SIRPB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | SIRPB2 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
SIRPB2 Polyclonal specifically detects SIRPB2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SIRPB2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
dJ776F14.2, Protein Tyrosine Phosphatase Non-Receptor Type Substrate 1-Like 3, Protein Tyrosine Phosphatase Non-Receptor Type Substrate Protein, Protein Tyrosine Phosphatase, Non-Receptor Type 1-Like, Protein Tyrosine Phosphatase, Non-Receptor Type Substrate 1-Like 3, PTPN1L, PTPNS1L3, Signal-Regulatory Protein Beta 2, Signal-Regulatory Protein Beta-2, SIRP-beta-2, SIRPG | |
SIRPB2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
Q5JXA9 | |
284759 | |
This antibody was developed against a recombinant protein corresponding to amino acids: GETLLLRCMVVGSCTDGMIKWVKVSTQDQQEIYNFKRGSFPGVMPMIQRTSEPLNCDYSIYIHNVTREHTGTYHCVRFDGLSEHSEMKSDEGTSVLVKGAGDPEPDLWIIQPQELVLGTTGDTVFLNCTV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title