Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SIRPB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545920
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545920 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545920 Supplier Invitrogen™ Supplier No. PA545920
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: VSSEDAGTYY CVKFQRKPNR QYLSGQGTSL KVKAKSTSSK EAEFTSEPAT Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SIRPB2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SIRPB2
Gene Accession No. Q5JXA9
Gene Alias dJ776F14.2; Protein tyrosine phosphatase non-receptor type substrate 1-like 3; protein tyrosine phosphatase non-receptor type substrate protein; protein tyrosine phosphatase, non-receptor type substrate 1-like 3; PTPN1L; PTPNS1L3; signal regulatory protein beta 2; signal-regulatory protein beta 2; signal-regulatory protein beta-2; SIRPB2; SIRP-beta-2
Gene Symbols SIRPB2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SIRPB2 (aa 232-281).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284759
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.