Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIRPD Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501735391 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-5391 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-5391 Supplier Proteintech Group Inc Supplier No. 265941AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Contains 1 Ig-like V-type (immunoglobulin-like) domain.

Specifications

Antigen SIRPD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene SIRPD
Gene Accession No. Q9H106
Gene Alias PTPNS1L2, SIRP delta, SIRPD
Gene Symbols SIRPD
Host Species Rabbit
Immunogen SIRPD Fusion Protein Ag24246
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 128646
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.