Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SIRT-Glo™ Assay and Screening Systems

Catalog No. PRG6452 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
100 mL
Product Type:
Screening System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRG6452 100 mL -
PRG6451 50 mL -
PRG6470 10 mL Screening System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRG6452 Supplier Promega Supplier No. G6452

Single-reagent-addition,homogeneous, luminescent assays that measure the relative activity of the NAD+-dependent histone deacetylase (HDAC) class III enzymes (sirtuins; SIRTs) from purified enzyme sources

  • Assay Advantages: simple measurement of deacetylating activities
  • Highly sensitive, fast data acquisition

Specifications

Quantity 100 mL
Content And Storage -20°C
For Use With (Application) Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling
Kit Contents 1 x 1 Vial SIRT-Glo substrate, 2 x 50μL developer reagent, 2 x 125mL SIRT-Glo Buffer
pH 8
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.